5-Cyclopentyl-1,3,4-oxadiazol-2-amine

Supplier: Matrix Scientific

037363-500MG
101843-208EA 290.77 USD
101843-208
5-Cyclopentyl-1,3,4-oxadiazol-2-amine
5-Cyclopentyl-1,3,4-oxadiazol-2-amine

Hazard: Irritant

Formula: C₇H₁₁N₃O
MDL Number: MFCD09806418
CAS Number: 90221-15-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR