1-(Methylsulfonyl)-1,4-diazepane hydrochloride

Supplier: Matrix Scientific

037360-500MG
101843-202EA 387.69 USD
101843-202
1-(Methylsulfonyl)-1,4-diazepane hydrochloride
1-(Methylsulfonyl)-1,4-diazepane hydrochloride

Hazard: Irritant

Formula: C₆H₁₅ClN₂O₂S
MDL Number: MFCD09376386
CAS Number: 550369-26-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR