[1-(Methylsulfonyl)piperidin-3-yl]methylamine hydrochloride

Supplier: Matrix Scientific

037359-500MG
101843-200EA 387.69 USD
101843-200
[1-(Methylsulfonyl)piperidin-3-yl]methylamine hydrochloride
[1-(Methylsulfonyl)piperidin-3-yl]methylamine hydrochloride

Hazard: Irritant

Formula: C₇H₁₇ClN₂O₂S
MDL Number: MFCD12027119
CAS Number: 869371-29-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR