1-Ethylpyrrolidine-2-carboxylic acid

Supplier: Matrix Scientific

037357-500MG
101843-196EA 260 USD
101843-196
1-Ethylpyrrolidine-2-carboxylic acid
1-Ethylpyrrolidine-2-carboxylic acid

Hazard: Irritant

Formula: C₇H₁₃NO₂
MDL Number: MFCD11505336
CAS Number: 165552-33-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR