4-(1H-Pyrazol-1-yl)butan-2-one

Supplier: Matrix Scientific

037354-500MG
101843-190EA 144.9 USD
101843-190
4-(1H-Pyrazol-1-yl)butan-2-one
4-(1H-Pyrazol-1-yl)butan-2-one

Hazard: Irritant

Formula: C₇H₁₀N₂O
MDL Number: MFCD00020740
CAS Number: 89943-03-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR