N-(tert-Butyl)-2-cyanoacetamide

Supplier: Matrix Scientific

037350-500MG
101843-182EA 193.85 USD
101843-182
N-(tert-Butyl)-2-cyanoacetamide
N-(tert-Butyl)-2-cyanoacetamide

Hazard: Irritant

Formula: C₇H₁₂N₂O
MDL Number: MFCD01351088
CAS Number: 108168-88-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR