2-Cyano-N-cyclohexylacetamide

Supplier: Matrix Scientific

037349-500MG
101843-180EA 146.15 USD
101843-180
2-Cyano-N-cyclohexylacetamide
2-Cyano-N-cyclohexylacetamide

Hazard: Irritant

Formula: C₉H₁₄N₂O
MDL Number: MFCD00228093
CAS Number: 15029-38-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR