2-Cyano-N-isopropylacetamide

Supplier: Matrix Scientific

037348-500MG
101843-178EA 86.25 USD
101843-178
2-Cyano-N-isopropylacetamide
2-Cyano-N-isopropylacetamide

Hazard: Irritant

Formula: C₆H₁₀N₂O
MDL Number: MFCD00828755
CAS Number: 52573-74-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR