N-Benzyl-2-cyanoacetamide

Supplier: Matrix Scientific

037347-500MG
101843-176EA 109.25 USD
101843-176
N-Benzyl-2-cyanoacetamide
N-Benzyl-2-cyanoacetamide

Hazard: Irritant

Formula: C₁₀H₁₀N₂O
MDL Number: MFCD00461336
CAS Number: 10412-93-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR