2-Cyano-N-hexylacetamide

Supplier: Matrix Scientific

037345-500MG
101843-170EA 146.15 USD
101843-170
2-Cyano-N-hexylacetamide
2-Cyano-N-hexylacetamide

Hazard: Irritant

Formula: C₉H₁₆N₂O
MDL Number: MFCD01349558
CAS Number: 52493-37-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR