3-Oxo-3-(1-pyrrolidinyl)propanenitrile

Supplier: Matrix Scientific

037344-500MG
101843-168EA 144.9 USD
101843-168
3-Oxo-3-(1-pyrrolidinyl)propanenitrile
3-Oxo-3-(1-pyrrolidinyl)propanenitrile

Hazard: Irritant

Formula: C₇H₁₀N₂O
MW: 138.17 g/mol
MDL Number: MFCD00020838
CAS Number: 14227-95-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR