(1-Allylpyrrolidin-3-yl)methanol

Supplier: Matrix Scientific

037342-500MG
101843-164EA 387.69 USD
101843-164
(1-Allylpyrrolidin-3-yl)methanol
(1-Allylpyrrolidin-3-yl)methanol

Hazard: Irritant

Formula: C₈H₁₅NO
MDL Number: MFCD09752217
CAS Number: 71548-32-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR