2-(Methylthio)-1,3-thiazole-4-carboxylic acid

Supplier: Matrix Scientific

Synonyms: 2-(Methylthio)thiazole-4-carboxylic acid

037341-500MG
101843-162EA 387.69 USD
101843-162
2-(Methylthio)-1,3-thiazole-4-carboxylic acid
2-(Methylthio)-1,3-thiazole-4-carboxylic acid

Hazard: Irritant

Formula: C₅H₅NO₂S₂
MW: 175.23 g/mol
MDL Number: MFCD11054031
CAS Number: 907543-75-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR