4-tert-Butyl-1,3-dihydro-2H-imidazol-2-one

Supplier: Matrix Scientific

037339-500MG
101843-158EA 387.69 USD
101843-158
4-tert-Butyl-1,3-dihydro-2H-imidazol-2-one
4-tert-Butyl-1,3-dihydro-2H-imidazol-2-one

Hazard: Irritant

Formula: C₇H₁₂N₂O
MDL Number: MFCD07357406
CAS Number: 623547-65-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR