4-Thien-2-ylpiperidine

Supplier: Matrix Scientific

Synonyms: 4-(2-Thienyl)piperidine2-(4-Piperidyl)thiophene

037335-500MG
101843-150EA 289.8 USD
101843-150
4-Thien-2-ylpiperidine
4-Thien-2-ylpiperidine

Hazard: Irritant

Formula: C₉H₁₃NS
MW: 167.27 g/mol
MDL Number: MFCD12027113
CAS Number: 198334-38-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR