N,N-Diethyl-4,4'-bipiperidine-1-sulfonamide

Supplier: Matrix Scientific

037334-500MG
101843-148EA 387.69 USD
101843-148
N,N-Diethyl-4,4'-bipiperidine-1-sulfonamide
N,N-Diethyl-4,4'-bipiperidine-1-sulfonamide

Hazard: Irritant

Formula: C₁₄H₂₉N₃O₂S
MDL Number: MFCD12027112
CAS Number: 1000941-37-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR