1-(Pyrrolidin-1-ylsulfonyl)-4,4'-bipiperidine
Supplier: Matrix Scientific
037333-500MG
101843-146EA
387.69
USD
101843-146
1-(Pyrrolidin-1-ylsulfonyl)-4,4'-bipiperidine
1-(Pyrrolidin-1-ylsulfonyl)-4,4'-bipiperidine
Hazard: Irritant
Formula:
C₁₄H₂₇N₃O₂S |
MDL Number:
MFCD12027111 CAS Number: 1000958-63-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...