3-(4-Fluorophenyl)furan-2,5-dione

Supplier: Matrix Scientific

037330-500MG
101843-140EA 217.35 USD
101843-140
3-(4-Fluorophenyl)furan-2,5-dione
3-(4-Fluorophenyl)furan-2,5-dione

Hazard: Irritant

Formula: C₁₀H₅FO₃
MDL Number: MFCD12027109
CAS Number: 58755-90-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR