N-Phenylpiperidin-4-amine

037328-500MG
101843-136EA 97.44 USD
101843-136
N-Phenylpiperidin-4-amine
N-Phenylpiperidin-4-amine

Hazard: Irritant

Formula: C₁₁H₁₆N₂
MDL Number: MFCD00198126
CAS Number: 23056-29-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR