N-(4-Methoxyphenyl)piperidin-4-amine

Supplier: Matrix Scientific

037325-500MG
101843-130EA 144.9 USD
101843-130
N-(4-Methoxyphenyl)piperidin-4-amine
N-(4-Methoxyphenyl)piperidin-4-amine

Hazard: Irritant

Formula: C₁₂H₁₈N₂O
MDL Number: MFCD06740344
CAS Number: 37656-54-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR