N-(Piperidin-3-ylmethyl)morpholine-4-carboxamide

Supplier: Matrix Scientific

037324-500MG
101843-128EA 289.8 USD
101843-128
N-(Piperidin-3-ylmethyl)morpholine-4-carboxamide
N-(Piperidin-3-ylmethyl)morpholine-4-carboxamide

Hazard: Irritant

Formula: C₁₁H₂₁N₃O₂
MDL Number: MFCD12027107
CAS Number: 1000941-33-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR