1-(2-Pyridin-4-ylethyl)piperidine-3-carboxylic acid
Supplier: Matrix Scientific
037317-500MG
101843-116EA
144.9
USD
101843-116
1-(2-Pyridin-4-ylethyl)piperidine-3-carboxylic acid
1-(2-Pyridin-4-ylethyl)piperidine-3-carboxylic acid
Hazard: Irritant
Formula:
C₁₃H₁₈N₂O₂ |
MDL Number:
MFCD06740685 CAS Number: 886506-27-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...