1-Acetyl-4,4'-bipiperidine

Supplier: Matrix Scientific

037314-500MG
101843-110EA 217.35 USD
101843-110
1-Acetyl-4,4'-bipiperidine
1-Acetyl-4,4'-bipiperidine

Hazard: Irritant

Formula: C₁₂H₂₂N₂O
MDL Number: MFCD06740337
CAS Number: 674774-74-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR