1-(7-Oxabicyclo[2.2.1]hept-2-yl)methanamine

Supplier: Matrix Scientific

037312-500MG
101843-106EA 461.54 USD
101843-106
1-(7-Oxabicyclo[2.2.1]hept-2-yl)methanamine
1-(7-Oxabicyclo[2.2.1]hept-2-yl)methanamine

Hazard: Irritant

Formula: C₇H₁₃NO
MDL Number: MFCD12027104
CAS Number: 153005-43-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR