N-(2-Piperidin-4-ylethyl)methanesulfonamide hydrochloride

Supplier: Matrix Scientific

037310-500MG
101843-102EA 384.62 USD
101843-102
N-(2-Piperidin-4-ylethyl)methanesulfonamide hydrochloride
N-(2-Piperidin-4-ylethyl)methanesulfonamide hydrochloride

Hazard: Irritant

Formula: C₈H₁₉ClN₂O₂S
MDL Number: MFCD08461904
CAS Number: 70922-37-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR