N,N-Dimethyl-4,4'-bipiperidine-1-sulfonamide

Supplier: Matrix Scientific

037309-500MG
101843-100EA 289.8 USD
101843-100
N,N-Dimethyl-4,4'-bipiperidine-1-sulfonamide
N,N-Dimethyl-4,4'-bipiperidine-1-sulfonamide

Hazard: Irritant

Formula: C₁₂H₂₅N₃O₂S
MDL Number: MFCD12027103
CAS Number: 1000958-59-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR