(3-Chlorophenyl)(4-piperazin-1-ylphenyl)methanone

Supplier: Matrix Scientific

037308-500MG
101843-098EA 217.35 USD
101843-098
(3-Chlorophenyl)(4-piperazin-1-ylphenyl)methanone
(3-Chlorophenyl)(4-piperazin-1-ylphenyl)methanone

Hazard: Irritant

Formula: C₁₇H₁₇ClN₂O
MDL Number: MFCD06740192
CAS Number: 886506-39-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR