4-(Piperidin-2-ylcarbonyl)morpholine

Supplier: Matrix Scientific

037304-500MG
101843-090EA 290.77 USD
101843-090
4-(Piperidin-2-ylcarbonyl)morpholine
4-(Piperidin-2-ylcarbonyl)morpholine

Hazard: Irritant

Formula: C₁₀H₁₈N₂O₂
MDL Number: MFCD06740217
CAS Number: 121791-04-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR