N-(4-Fluorophenyl)piperidin-4-amine

Supplier: Matrix Scientific

037303-500MG
101843-088EA 201.25 USD
101843-088
N-(4-Fluorophenyl)piperidin-4-amine
N-(4-Fluorophenyl)piperidin-4-amine

Hazard: Irritant

Formula: C₁₁H₁₅FN₂
MDL Number: MFCD06619353
CAS Number: 38043-08-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR