3-(4-Methoxyphenyl)piperidine

Supplier: Matrix Scientific

037300-500MG
101843-082EA 443.08 USD
101843-082
3-(4-Methoxyphenyl)piperidine
3-(4-Methoxyphenyl)piperidine

Hazard: Irritant

Formula: C₁₂H₁₇NO
MDL Number: MFCD09837353
CAS Number: 19725-26-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR