[1-(2-Furylmethyl)piperidin-4-yl]methylamine
Supplier: Matrix Scientific
037299-500MG
101843-080EA
592.31
USD
101843-080
[1-(2-Furylmethyl)piperidin-4-yl]methylamine
[1-(2-Furylmethyl)piperidin-4-yl]methylamine
Hazard: Irritant/Stored under Argon
Formula:
C₁₁H₁₈N₂O |
MDL Number:
MFCD09837352 CAS Number: 725211-89-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...