Sodium-1-(2-pyridin-4-ylethyl)piperidine-4-carboxylate

Supplier: Matrix Scientific

037297-500MG
101843-076EA 193.85 USD
101843-076
Sodium-1-(2-pyridin-4-ylethyl)piperidine-4-carboxylate
Sodium-1-(2-pyridin-4-ylethyl)piperidine-4-carboxylate

Hazard: Irritant

Formula: C₁₃H₁₇N₂NaO₂
MW: 256.28 g/mol
MDL Number: MFCD09759185
CAS Number: 1158260-61-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR