Methyl (4-fluorophenyl)(4-phenylpiperazin-1-yl)acetate hydrobromide

Supplier: Matrix Scientific

037295-500MG
101843-072EA 193.85 USD
101843-072
Methyl (4-fluorophenyl)(4-phenylpiperazin-1-yl)acetate hydrobromide
Methyl (4-fluorophenyl)(4-phenylpiperazin-1-yl)acetate hydrobromide

Hazard: Irritant

Formula: C₁₉H₂₂BrFN₂O₂
MDL Number: MFCD09837350
CAS Number: 1063629-70-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR