N-(4-Methoxyphenyl)pyrrolidin-3-amine

Supplier: Matrix Scientific

037292-500MG
101843-066EA 217.35 USD
101843-066
N-(4-Methoxyphenyl)pyrrolidin-3-amine
N-(4-Methoxyphenyl)pyrrolidin-3-amine

Hazard: Irritant

Formula: C₁₁H₁₆N₂O
MDL Number: MFCD06740349
CAS Number: 886506-09-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR