[1-(3-Phenylpropyl)piperidin-4-yl]methylamine

Supplier: Matrix Scientific

037291-500MG
101843-064EA 193.85 USD
101843-064
[1-(3-Phenylpropyl)piperidin-4-yl]methylamine
[1-(3-Phenylpropyl)piperidin-4-yl]methylamine

Hazard: Irritant

Formula: C₁₅H₂₄N₂
MDL Number: MFCD08059669
CAS Number: 223567-74-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR