(4-Ethoxyphenyl)(piperidin-4-yl)methanone hydrochloride
Supplier: Matrix Scientific
037287-500MG
101843-056EA
461.54
USD
101843-056
(4-Ethoxyphenyl)(piperidin-4-yl)methanone hydrochloride
(4-Ethoxyphenyl)(piperidin-4-yl)methanone hydrochloride
Hazard: Irritant
Formula:
C₁₄H₂₀ClNO₂ |
MDL Number:
MFCD08059667 CAS Number: 1158249-49-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...