[1-(4-Fluorobenzyl)piperidin-4-yl]methylamine

Supplier: Matrix Scientific

037286-500MG
101843-054EA 290.77 USD
101843-054
[1-(4-Fluorobenzyl)piperidin-4-yl]methylamine
[1-(4-Fluorobenzyl)piperidin-4-yl]methylamine

Hazard: Irritant

Formula: C₁₃H₁₉FN₂
MDL Number: MFCD06809642
CAS Number: 174561-02-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR