4-[(4-Fluorophenyl)amino]piperidine-4-carboxamide

Supplier: Matrix Scientific

037282-500MG
101843-046EA 387.69 USD
101843-046
4-[(4-Fluorophenyl)amino]piperidine-4-carboxamide
4-[(4-Fluorophenyl)amino]piperidine-4-carboxamide

Hazard: Irritant

Formula: C₁₂H₁₆FN₃O
MDL Number: MFCD07021598
CAS Number: 878441-19-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR