2-Methyl-4-piperazin-1-yl-6-(trifluoromethyl)pyrimidine

Supplier: Matrix Scientific

037278-500MG
101843-038EA 230.77 USD
101843-038
2-Methyl-4-piperazin-1-yl-6-(trifluoromethyl)pyrimidine
2-Methyl-4-piperazin-1-yl-6-(trifluoromethyl)pyrimidine

Hazard: Irritant

Formula: C₁₀H₁₃F₃N₄
MW: 246.24 g/mol
MDL Number: MFCD07365354
CAS Number: 893752-50-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR