1-(5-Nitropyridin-2-yl)piperidin-4-amine

Supplier: Matrix Scientific

037275-500MG
101843-032EA 290.77 USD
101843-032
1-(5-Nitropyridin-2-yl)piperidin-4-amine
1-(5-Nitropyridin-2-yl)piperidin-4-amine

Hazard: Irritant

Formula: C₁₀H₁₄N₄O₂
MDL Number: MFCD06619062
CAS Number: 252577-85-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR