(3-Pyridin-3-yl-1,2,4-oxadiazol-5-yl)methanol

Supplier: Matrix Scientific

037274-500MG
101843-030EA 387.69 USD
101843-030
(3-Pyridin-3-yl-1,2,4-oxadiazol-5-yl)methanol
(3-Pyridin-3-yl-1,2,4-oxadiazol-5-yl)methanol

Hazard: Irritant

Formula: C₈H₇N₃O₂
MDL Number: MFCD12027101
CAS Number: 1119451-10-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR