N-Methyl-N-[(3-methyl-1,2,4-oxadiazol-5-yl)methyl]amine

Supplier: Matrix Scientific

037272-500MG
101843-026EA 387.69 USD
101843-026
N-Methyl-N-[(3-methyl-1,2,4-oxadiazol-5-yl)methyl]amine
N-Methyl-N-[(3-methyl-1,2,4-oxadiazol-5-yl)methyl]amine

Hazard: Irritant

Formula: C₅H₉N₃O
MW: 127.15 g/mol
MDL Number: MFCD11124699
CAS Number: 933683-06-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR