N-[(3-Cyclopropyl-1,2,4-oxadiazol-5-yl)methyl]-N-methylamine

Supplier: Matrix Scientific

037270-500MG
101843-022EA 290.77 USD
101843-022
N-[(3-Cyclopropyl-1,2,4-oxadiazol-5-yl)methyl]-N-methylamine
N-[(3-Cyclopropyl-1,2,4-oxadiazol-5-yl)methyl]-N-methylamine

Hazard: Irritant

Formula: C₇H₁₁N₃O
MDL Number: MFCD11116694
CAS Number: 1036471-29-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR