(3-tert-Butyl-1,2,4-oxadiazol-5-yl)methylamine hydrochloride

Supplier: Matrix Scientific

037269-500MG
101843-020EA 362.25 USD
101843-020
(3-tert-Butyl-1,2,4-oxadiazol-5-yl)methylamine hydrochloride
(3-tert-Butyl-1,2,4-oxadiazol-5-yl)methylamine hydrochloride

Hazard: Irritant

Formula: C₇H₁₄ClN₃O
MDL Number: MFCD12027099
CAS Number: 944901-66-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR