[3-(3-Methylphenyl)-1,2,4-oxadiazol-5-yl]methylamine hydrochloride

Supplier: Matrix Scientific

037266-500MG
101843-014EA 217.35 USD
101843-014
[3-(3-Methylphenyl)-1,2,4-oxadiazol-5-yl]methylamine hydrochloride
[3-(3-Methylphenyl)-1,2,4-oxadiazol-5-yl]methylamine hydrochloride

Hazard: Irritant

Formula: C₁₀H₁₂ClN₃O
MDL Number: MFCD12027096
CAS Number: 890324-13-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR