N,N-Dimethyl-2-(methylamino)propanamide

Supplier: Matrix Scientific

037264-500MG
101843-010EA 290.77 USD
101843-010
N,N-Dimethyl-2-(methylamino)propanamide
N,N-Dimethyl-2-(methylamino)propanamide

Hazard: Irritant

Formula: C₆H₁₄N₂O
MDL Number: MFCD09880870
CAS Number: 69902-16-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR