2-Amino-N,N-dimethylpropanamide hydrochloride

Supplier: Matrix Scientific

037263-500MG
101843-008EA 387.69 USD
101843-008
2-Amino-N,N-dimethylpropanamide hydrochloride
2-Amino-N,N-dimethylpropanamide hydrochloride

Hazard: Irritant

Formula: C₅H₁₃ClN₂O
MDL Number: MFCD12027094
CAS Number: 124491-96-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR