[(2-Methoxyethyl)(methyl)amino]acetic acid

Supplier: Matrix Scientific

037259-500MG
101843-002EA 387.69 USD
101843-002
[(2-Methoxyethyl)(methyl)amino]acetic acid
[(2-Methoxyethyl)(methyl)amino]acetic acid

Hazard: Irritant

Formula: C₆H₁₃NO₃
MDL Number: MFCD08691616
CAS Number: 915925-21-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR