2-Pyrrolidin-1-ylpropan-1-ol

Supplier: Matrix Scientific

037258-500MG
101843-000EA 387.69 USD
101843-000
2-Pyrrolidin-1-ylpropan-1-ol
2-Pyrrolidin-1-ylpropan-1-ol

Hazard: Irritant

Formula: C₇H₁₅NO
MDL Number: MFCD12027091
CAS Number: 53663-19-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR