N-Methyl-2-(methylamino)propanamide

Supplier: Matrix Scientific

037252-500MG
101842-988EA 230.77 USD
101842-988
N-Methyl-2-(methylamino)propanamide
N-Methyl-2-(methylamino)propanamide

Hazard: Irritant

Formula: C₅H₁₂N₂O
MDL Number: MFCD11057910
CAS Number: 63095-84-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR